5-Bromo-2-hydroxy-3-picoline
Glass bottle for solids, 25 g
Catalog # COBBPY-1118-25G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one
- CAS Number:89488-30-2
- Formula:C₆H₆BrNO
- MDL Number:MFCD03427657
- Molecular Weight:188.02 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



