0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Bis(pinacolato)diboron
Bis(pinacolato)diboron
Bis(pinacolato)diboron
Bis(pinacolato)diboron
Glass bottle for solids, 100 g
COBBBB-2214-100G
:
COMBI-BLOCKS
Bis(pinacolato)diboron
COBBBB-2214-100G
:
COMBI-BLOCKS
:
73183-34-3
Expand image
Bis(pinacolato)diboron
Glass bottle for solids, 100 g
Add to cart
:
73183-34-3
Unit of Measure
1 * 100 g
-
+
:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
:
73183-34-3
:
C₁₂H₂₄B₂O₄
:
MFCD00799570
:
253.94 g/mol
Documents
Pack type:
Glass bottle for solids
Pk:
100 g