Bis(pinacolato)diboron
Glass bottle for solids, 25 g
Catalog # COBBBB-2214-25G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
- CAS Number:73183-34-3
- Formula:C₁₂H₂₄B₂O₄
- MDL Number:MFCD00799570
- Molecular Weight:253.94 g/mol
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



