5-Bromo-2-chlorophenol
Glass bottle for solids, 25 g
Catalog # COBBOS-7612-25G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:183802-98-4
- Formula:C₆H₄BrClO
- MDL Number:MFCD00672940
- Molecular Weight:207.45 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



