Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Bromobenzonitrile

Glass bottle for solids, 25 g
Catalog # COBBBK-4527-25G
Supplier:   COMBI-BLOCKS
4-Bromobenzonitrile
Catalog # COBBBK-4527-25G
Supplier:  COMBI-BLOCKS
CAS Number:  623-00-7
4-Bromobenzonitrile

Glass bottle for solids, 25 g

CAS Number:  623-00-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  p-Bromobenzonitrile
  • CAS Number:623-00-7
  • Formula:C₇H₄BrN
  • MDL Number:MFCD00001811
  • Molecular Weight:182.02 g/mol
  • Boiling Pt:236 °C (1013 hPa)
  • Melting Pt:∼90 °C
  • Density:1.562 g/cm³ (20 °C)
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
  • EINECS:210-764-9
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Cat. No.:
    COBBBK-4527-25G

Specifications