ContactUsLinkComponent
Safety cabinet with drawer: Outer carcass: 888 x 595 x 635 (WxDxH) mm, Inner carcass: 791 x 463 x 480 (WxDxH) mm, 1 drawer Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, drawer in RAL 7035 light grey, Inner carcass in RAL 7035 ligh 1 * 1 items
Catalog # 132-0641
Supplier:  DUPERTHAL
undefined
Safety cabinet with drawer: Outer carcass: 888 x 595 x 635 (WxDxH) mm, Inner carcass: 791 x 463 x 480 (WxDxH) mm, 1 drawer Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, drawer in RAL 7035 light grey, Inner carcass in RAL 7035 ligh 1 * 1 items
Catalog # 132-0641
Supplier:  DUPERTHAL