Antigen Retrieval Buffer (100X Citrate Buffer), Abcam
Product Details & Documents
Antigen retrieval 100X citrate buffer (pH 5.1 - 5.5) is a special solution that enables rehydration and antigen retrieval to be performed in formalin-fixed, paraffin-embedded tissue sections with one step. - Cited in over 25 publications - Contains citrate buffer, 1.5% 2-butoxyethanol, 1% hydrochloric acid - Optimized for citrate buffer-treated antibodies - Contains detergent for paraffin emulsification
- pH: 5.1 - 5.5 Constituents: Citrate Buffer, 1.5% 2-butoxyethanol, 1% Hydrochloric acid
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:Antigen Retrieval Buffer (100X Citrate Buffer)
- Cat. No.:ABCAAB93678-125
- Storage temperature:Store at Ambient.



