Orders LinkContactUsLinkComponent
dNTP sets (dA, dC, dG, dT) 100 mM solutions
dNTP sets (dA, dC, dG, dT) 100 mM solutions
dNTP set of four (dA, dC, dG, dT) 100 mM solutions, 4×0,25 ml, 1 SET
Catalog # 786-460
Supplier:   G-Biosciences
dNTP sets (dA, dC, dG, dT) 100 mM solutions
Catalog # 786-460
Supplier:  G-Biosciences
CAS Number:  
dNTP sets (dA, dC, dG, dT) 100 mM solutions
dNTP sets (dA, dC, dG, dT) 100 mM solutions
dNTP set of four (dA, dC, dG, dT) 100 mM solutions, 4×0,25 ml, 1 SET
CAS Number:  
Price
197,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Set consists of 100 mM aqueous solutions of dATP, dCTP, dGTP and dTTP each supplied in a separate vial. Also available separately.
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    dNTP set of four (dA, dC, dG, dT) 100 mM solutions, 4×0,25 ml
Specifications