Orders LinkContactUsLinkComponent
Sample clean up, Perfect-FOCUS™
Sample clean up, Perfect-FOCUS™
Perfect-FOCUS™, 6 preps, 1 KIT
Catalog # 786-124T
Supplier:   G-Biosciences
Sample clean up, Perfect-FOCUS™
Catalog # 786-124T
Supplier:  G-Biosciences
CAS Number:  
Sample clean up, Perfect-FOCUS™
Sample clean up, Perfect-FOCUS™
Perfect-FOCUS™, 6 preps, 1 KIT
CAS Number:  
Price
64,10 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Perfect-FOCUS™ is based on quantitative precipitation and concentration of protein solutions using UPPA™ (Universal Protein Precipitation Agent) and has been specifically developed for preparing lower conductivity protein samples for iso-electric focusing gels. Perfect-FOCUS™ is designed to clean and remove interfering agents from protein samples and concentrate protein solutions that give poor protein spot resolution during 2D electrophoresis.
Protein solutions as dilute as 1 ng/ml, if treated with Perfect-FOCUS™, can be quantitatively precipitated into a small volume. Protein precipitation is not affected by the presence of detergents or other common laboratory agents.
The collected protein sample has a conductivity of approximately 40 to 50 µS and is substantially free from non-protein agents, making the protein sample ideal for critical IEF and 2D gel electrophoresis studies, with improved spot resolution and greatly reduced spot streaking. Perfect-FOCUS™ treated protein samples are compatible with mass spectrometry analysis and show identical peptide spectra.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Perfect-FOCUS™, 6 preps
Specifications