Orders LinkContactUsLinkComponent
Protein blocking buffers, Superior™
Protein blocking buffers, Superior™
Superior™ blocking buffer in TBS, 500 mL
Catalog # 786-658
Supplier:   G-Biosciences
Protein blocking buffers, Superior™
Catalog # 786-658
Supplier:  G-Biosciences
CAS Number:  
Protein blocking buffers, Superior™
Protein blocking buffers, Superior™
Superior™ blocking buffer in TBS, 500 mL
CAS Number:  
Price
148,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Blocking buffer is an enhanced blocking agent available in multiple formats. Superior™ Blocking Buffer contains a proprietary antigenically non-determinant protein for blocking Western blot membranes, blocking and storage of ELISA plates, and blocking prior to immunohistochemistry staining.
Ideal for a high signal to background ratio in most systems and has a rapid blocking time of about two minutes for ELISA. It uses a non-serum protein and does not contain biotin or other animal source proteins that interfere with immuno-complexes. It is also suitable for assays that use avidin/streptavidin systems.
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Superior™ blocking buffer in TBS
Specifications