Order Entry
Export
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ SCX

Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 75 ml, 10000 mg
Catalog # HYPE60108-718
Solid phase extraction cartridges, HyperSep™ SCX
Catalog # HYPE60108-718
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 75 ml, 10000 mg

Price
454,00 €
Log in for availability
1 * 10 items
Product Details & Documents

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    75 ml
  • Seq:
    09
  • Bed weight:
    10000 mg
  • Phase:
    SCX
  • Cat. No.:
    HYPE60108-718