Order Entry
Export
Orders LinkContactUsLinkComponent
Connectors, for GL threaded connections

Connectors, for GL threaded connections

GL 14 screw caps, 8 mm
Catalog # BOHLD584-14
Supplier:   Bohlender
Connectors, for GL threaded connections
Catalog # BOHLD584-14
Supplier:  Bohlender
Connectors, for GL threaded connections
Connectors, for GL threaded connections

GL 14 screw caps, 8 mm

Price
8,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Connector made from PP with elastic sealing lip and FPM O-ring

  • Good chemical resistance
  • Temperature resistant from −20 to +110°C
  • Suitable for vacuum applications

For use with GL screw caps.

Specifications
  • For:
    GL 14 screw caps
  • Tubing int.Ø:
    8 mm