Order Entry
Export
Orders LinkContactUsLinkComponent
Multiple distributors for bottles , BOLA

Multiple distributors for bottles , BOLA

GL 32, 3 × max. 8 mm, 3 × GL 14
Catalog # BOHLD621-08
Supplier:   Bohlender
Multiple distributors for bottles , BOLA
Catalog # BOHLD621-08
Supplier:  Bohlender
Multiple distributors for bottles , BOLA
Multiple distributors for bottles , BOLA

GL 32, 3 × max. 8 mm, 3 × GL 14

Price
93,90 €
Log in for availability
1 * 1 items
Product Details & Documents

PTFE.

  • Very good chemical resistance
  • Temperature resistant from -200 to +250 °C
  • Suitable for vacuum applications

For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with an external diameter of max. 8 mm can be inserted through the stopcocks.

Specifications
  • Thread:
    GL 32
  • Tube ext.Ø:
    3 × max. 8 mm
  • Neck:
    3 × GL 14