Orders LinkContactUsLinkComponent
Non return valves, BOLA
Non return valves, BOLA
Non return valve with thread of fitting M14 x 2, for tubing O-Ø 4 mm, total length 110 mm, PTFE
Catalog # BOHLE680-21
Supplier:   Bohlender
Non return valves, BOLA
Catalog # BOHLE680-21
Supplier:  Bohlender
Non return valves, BOLA
Non return valves, BOLA
Non return valve with thread of fitting M14 x 2, for tubing O-Ø 4 mm, total length 110 mm, PTFE
Price
371,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These non return valves are made from PTFE, with nuts made from glass fibre reinforced PTFE for connecting tubing or tubes.
  • Universally chemically resistant
  • Flow medium comes into contact with PTFE or PFA only
The built-in lock function only allows flow in one direction, the flow direction is marked by an arrow.
Specifications
  • Description:
    Non return valve with thread of fitting M14 x 2, for tubing O-Ø 4 mm, total length 110 mm
  • Material:
    PTFE
  • Seq:
    01