Orders LinkContactUsLinkComponent
Control valves with GL thread, BOLA
GL control valve with two-way straight bore Ø 4 mm, thread GL 14, PTFE
Catalog # BOHLE694-14
Supplier:   Bohlender
Control valves with GL thread, BOLA
Catalog # BOHLE694-14
Supplier:  Bohlender
Control valves with GL thread, BOLA
Control valves with GL thread, BOLA
GL control valve with two-way straight bore Ø 4 mm, thread GL 14, PTFE
Price
535,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Two-way valve with straight bore and two connections with GL thread completely made of PTFE.
  • Suitable for vacuum
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Applications include for distributing liquids or gases, manual regulation for constant flow.
Specifications
  • Description:
    GL control valve with two-way straight bore Ø 4 mm, thread GL 14, PTFE