Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Protein Purification
Protein Purification Equipment
Accessories for chromatography systems
Accessories for chromatography systems
Print
Accessories for chromatography systems
Termination plug, 1
Catalog # 18-1118-59
Supplier:
Cytiva
Accessories for chromatography systems
Catalog # 18-1118-59
Supplier:
Cytiva
CAS Number:
Expand image
10+
Return to Top
Accessories for chromatography systems
Termination plug, 1
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
23,70 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Datasheet
18100368_DATA FILE
18100368_DATA FILE 1
18102712_DATA FILE
18102712_DATA FILE 1
18321603_DATA FILE
Manual
18102712_HAND BOOK
18321603-HAND BOOK
18321603_INSTRUCTIONS
Additional Product Details
Cytiva CyDye Licensing Statements
Cytiva General Licensing Statements
Specifications
Specifications
Pk:
1
Description:
Termination plug
Specifications