Order Entry
Export
Orders LinkContactUsLinkComponent
G418 disulphate in solution

G418 disulphate in solution

Glass bottle, 50 mL
Catalog # A6798.0050
Supplier:   PanReac AppliChem
G418 disulphate in solution
Catalog # A6798.0050
Supplier:  PanReac AppliChem
CAS Number:  108321-42-2
G418 disulphate in solution

Glass bottle, 50 mL

CAS Number:  108321-42-2
Price
337,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

G418 disulphate supplied as a ready-to-use sterile solution.

Concentration: 50 mg/ml

Synonyms:  G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
  • CAS Number:108321-42-2
  • Formula:C₂₀H₄₄N₄O₁₈S₂
  • MDL Number:MFCD00058314
  • Molecular Weight:692.72 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications