Orders LinkContactUsLinkComponent
Inoculating loops and needles
Inoculating loops and needles
Innoculating loops, 1 µl, White, 48/peel pack
Catalog # SARS86.1567.050
Supplier:   SARSTEDT
Inoculating loops and needles
Catalog # SARS86.1567.050
Supplier:  SARSTEDT
Inoculating loops and needles
Innoculating loops, 1 µl, White, 48/peel pack
Price
83,90 €
Log in for availability
1 * 1.920 items
Product Details & Documents
PS, gamma sterilised. Time-saving, safe and easy to use. For laboratories with high workloads.
Specifications
  • Packed:
    48/peel pack
  • Capacity:
    1 µl
  • Colour:
    White
  • Description:
    Innoculating loops