Replacement Erlenmeyer flask caps, Corning®
Vent cap, 43 mm, for 500 ml and 1 l flask (made to order only, 5 case minimum)
Catalog # 392-0292
Supplier: Corning
Jump to:
Product Details & Documents
Made of PP.
Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
- Description:Vent cap, 43 mm, for 500 ml and 1 l flask (made to order only, 5 case minimum)
- Seq:01