Dry box gloves
Surgical-type latex hands, for use with sleeves, 7, White, 0,009 gauge
Catalog # LBCP5006200
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Glove size:7
- Colour:White
- Thickness:0,009 gauge
- Model:Surgical-type latex hands, for use with sleeves