Inoculating loops and needles
1 µl, Natural
Supplier: KARTELL
Jump to:
Product Details & Documents
High impact PS, sterile.
- Choice of either 1 µl or 10 µl loop on one end and needle at the other
- Sterilised by gamma irradiation
Packaging: 20 per bag.
Documents
Specifications
- Capacity:1 µl
- Colour:Natural
- Seq:01