Disposable earplugs, E-A-RSoft™
Earplug, roll down, with blue cord, Yellow
Catalog # 111-1797
Supplier: 3M
Jump to:
Product Details & Documents
E-A-RSoft™ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexibility and a good seal with optimum comfort. Once inserted in the ear, the foam plugs expand to provide a snug and secure custom fit. These roll down earplugs are available with and without cord and be used with the One Touch™ dispenser.
- Soft hypoallergenic foam material for lower pressure inside the ear
- Smooth, dirt resistant surface for better hygiene, durability and comfort
- Polyester cord helps prevent hearing loss
- One size that fits most ear canals
SNR: 37 dB
Certifications: EN 352-2
Specifications
- Colour:Yellow
- Description:Earplug, roll down, with blue cord
- Seq:02
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Earplugs, E-A-RSoft™ Yellow Neons™, 34 dB, refill bottle for dispenserEarplugs, E-A-RSoft™ Yellow Neons™, 34 dB, refill bottle for dispenser111-1585YellowLog in for availability1 * 500 PAIR - 105,00 €
Product Information
- Catalog #ColourAvailabilityPrice
- Earplugs E-A-R Soft™ - Refill box for dispenser bottleEarplugs E-A-R Soft™ - Refill box for dispenser bottle111-2422YellowLog in for availability1 * 500 items - 94,50 €



