Order Entry
Export
Orders LinkContactUsLinkComponent
Mortars without Pestles, Borosilicate Glass

Mortars without Pestles, Borosilicate Glass

225 ml, 100 mm
Catalog # HECH47028010
Mortars without Pestles, Borosilicate Glass
Catalog # HECH47028010
Mortars without Pestles, Borosilicate Glass
Mortars without Pestles, Borosilicate Glass

225 ml, 100 mm

Price
26,80 €
Log in for availability
1 * 1 items
Product Details & Documents

Borosilicate glass, with spout and rough interior surface.

Specifications
  • Ø:
    100 mm
  • Capacity:
    225 ml
  • Seq:
    02