Orders LinkContactUsLinkComponent
Immersion coolers
Immersion coolers
140 mm, 29/32 NS
  WITG1.786.229.S
Immersion coolers
  WITG1.786.229.S
Immersion coolers
Immersion coolers
140 mm, 29/32 NS
69,00 €
1 * 1 items
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it. Water connection with two glass olives.
  • Cone size:
    29/32 NS
  • Jacket length:
    140 mm