ContactUsLinkComponent
Pumphead, material PTFE, flowrate minimum 34ml/min, maximum 99 degree celsius, carbon, differential pressure 4.5 bar maximum, 316 st ainless steel. 1 * 1 items
Catalog # ISMCFMI017
Pumphead, material PTFE, flowrate minimum 34ml/min, maximum 99 degree celsius, carbon, differential pressure 4.5 bar maximum, 316 st ainless steel. 1 * 1 items
Catalog # ISMCFMI017