Accessories for PURELAB Flex Systems
Biofilter 0,2 µ RNAse free: removes pyrogens and endotoxins for molecular grade water applications.
For PURELAB® flex 2 and 3.
For PURELAB® flex 2 and 3.
Catalog # 171-0365
Supplier: ELGA LabWater
Jump to:
Product Details & Documents
Specifications
- Description:Biofilter 0,2 µ RNAse free: removes pyrogens and endotoxins for molecular grade water applications.
For PURELAB® flex 2 and 3. - Seq:02