Orders LinkContactUsLinkComponent
Accessories for PURELAB Flex Systems
Accessories for PURELAB Flex Systems
Leak detector: prevents water damage by automatically turning off the water supply to the flex system when the sensor detects water. Includes battery, tap sieve, fittings and user manual.
For PURELAB® flex 2 and 3
Catalog # 171-0355
Accessories for PURELAB Flex Systems
Catalog # 171-0355
Supplier:  
Accessories for PURELAB Flex Systems
Leak detector: prevents water damage by automatically turning off the water supply to the flex system when the sensor detects water. Includes battery, tap sieve, fittings and user manual.
For PURELAB® flex 2 and 3
Discontinued
Product Details & Documents
Specifications
  • Description:
    Leak detector: prevents water damage by automatically turning off the water supply to the flex system when the sensor detects water. Includes battery, tap sieve, fittings and user manual.
    For PURELAB® flex 2 and 3
  • Seq:
    15