Orders LinkContactUsLinkComponent
Accessories for PURELAB Flex Systems
Accessories for PURELAB Flex Systems
Preconditioning cartridge holder: to ensure optimum performance when feedwater is > 1 µS/cm
For PURELAB® flex 2 only
Catalog # 171-0353
Supplier:   ELGA LabWater
Accessories for PURELAB Flex Systems
Catalog # 171-0353
Supplier:  ELGA LabWater
Accessories for PURELAB Flex Systems
Preconditioning cartridge holder: to ensure optimum performance when feedwater is > 1 µS/cm
For PURELAB® flex 2 only
Price
662,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Preconditioning cartridge holder: to ensure optimum performance when feedwater is > 1 µS/cm
    For PURELAB® flex 2 only