Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels, with short stem, PP

Funnels, with short stem, PP

110 mm, 14 mm, 150 mm
Catalog # SEMA0221
Supplier:   SEMADENI
Funnels, with short stem, PP
Catalog # SEMA0221
Supplier:  SEMADENI
Funnels, with short stem, PP
Funnels, with short stem, PP

110 mm, 14 mm, 150 mm

Price
20,30 €
Log in for availability
1 * 1 items
Product Details & Documents

Funnels, made of PP, translucent, with slim stem.

  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable
Specifications
  • Funnel O-Ø:
    150 mm
  • Stem O-Ø:
    14 mm
  • Stem length:
    110 mm