Push-in caps for tubes
LDPE, Caps, Ø 19 mm, Natural
Catalog # SEMA1467
Supplier: SEMADENI
Jump to:
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
- Colour:Natural
- Description:Caps, Ø 19 mm
- Material:LDPE
- Seq:17