Telescopic handles
Lightweight handle, extendable, with blue clip, 1000 - 1800 mm
Catalog # 129-0456
Supplier: FHP VILEDA SCS
Jump to:
Product Details & Documents
These lightweight, strong extendable handles are perfect for all controlled environments. Fully autoclavable.
- Extremely easy to use and extend with a simple twist of the wrist
- Resist autoclave, gamma and ETO processes
- Ergonomic center and anti-slip grips
- Available with colour coding clips (Blue, Red, Yellow, Green)
For use with all Vileda Professional frames and CE Squeegee.
Specifications
- Description:Lightweight handle, extendable, with blue clip
- Length:1000 - 1800 mm
- Seq:02