Orders LinkContactUsLinkComponent
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
1500 ml, 45/40 NS, 29/32 NS
Catalog # WITG0.851.150
Supplier:   witeg Labortechnik
Sulfonation flasks, with three necks
Catalog # WITG0.851.150
Supplier:  witeg Labortechnik
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
1500 ml, 45/40 NS, 29/32 NS
Price
212,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifications
  • Capacity:
    1500 ml
  • Seq:
    05
  • Side necks:
    29/32 NS
  • Centre neck:
    45/40 NS