Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for pH electrodes
Accessories for pH electrodes
Print
Accessories for pH electrodes
pH electrode cleaning solution, 500 ml
Catalog # JENW16156-77
Supplier:
Cole-Parmer
Accessories for pH electrodes
Catalog # JENW16156-77
Supplier:
Cole-Parmer
Expand image
Return to Top
Accessories for pH electrodes
pH electrode cleaning solution, 500 ml
Specifications
Add to cart
Jump to:
Specifications
Price
70,60 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Description:
pH electrode cleaning solution, 500 ml