Orders LinkContactUsLinkComponent
Screw caps for tubes
Screw caps for tubes
PP, 18-415
Catalog # DELT617300
Supplier:   DELTALAB
Screw caps for tubes
Catalog # DELT617300
Supplier:  DELTALAB
Screw caps for tubes
PP, 18-415
Price
284,00 €
Log in for availability
1 * 500 items
Product Details & Documents
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
  • Both models of caps are fully autoclavable and are pharmaceutical quality
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Documents
Specifications
  • Thread:
    18-415
  • Material:
    PP