Orders LinkContactUsLinkComponent
Cork borer sets
Cork borer, 13,8 mm
Catalog # 217-0838
Supplier:   USBECK Laborgeräte
Cork borer sets
Catalog # 217-0838
Cork borer sets
Cork borer sets
Cork borer, 13,8 mm
Price
10,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Brass, nickel plated, with handles and ejector.
  • For manual boring of cork plugs
  • Easy to clean
  • Also available, cork drill sharpener compatible with all sets
Specifications
  • Hole Ø:
    13,8 mm
  • Description:
    Cork borer
  • Seq:
    11
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cork borer sharpener
    Cork borer sharpener
    217-9692
    Log in for availability
    1 * 1 items - 28,70 €