Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
#1,5, 18 mm
Catalog # HECH92101401120
Cover glasses
Catalog # HECH92101401120
Cover glasses
Cover glasses
#1,5, 18 mm
Price
249,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
High spectral transmission, excellent flatness, optimum resistance to chemicals.
  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks
Certifications: ISO 8255
Packaging: In tight-closing plastic boxes for protection against damp and climatic influences.
Specifications
  • Ø:
    18 mm
  • No.:
    #1,5
  • Seq:
    01