Order Entry
Export
Orders LinkContactUsLinkComponent
Technical pH buffer solutions, HI 5000 series

Technical pH buffer solutions, HI 5000 series

4,01, ±0,01, 25 °C, 500 mL
Catalog # HANNHI5004
Supplier:   Hanna
Technical pH buffer solutions, HI 5000 series
Catalog # HANNHI5004
Supplier:  Hanna
CAS Number:  
Technical pH buffer solutions, HI 5000 series

4,01, ±0,01, 25 °C, 500 mL

CAS Number:  
Price
70,30 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • Colour coded labels
  • ±0,01 pH

Certifications: Technical buffer solutions with certificate.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,01
  • pH value:
    4,01

Specifications