PIG® Burpless® Poly Drum Funnel
Funnel, red/black, 560×290 mm
Catalog # NEWPDRM138-RD
Supplier: NEW PIG
Jump to:
Product Details & Documents
Burpless design vents during pouring for fast, smooth flow with no dangerous splashback.
- Lets you pour without venting through 3/4" bung
- Thick-wall, 100% polyethylene construction resists UV rays, rust, corrosion and most chemicals
- Designed for use with non-hazardous liquids: coolants, non-flammable oils and water-based liquids
- Lockable to prevent unauthorised access
- Multiple lid colour options let you segregate drums
- Attached hinged lid won't get lost or separated from funnel
- Ridged interior creates a level draining surface for containers and promotes faster draining
Built-in debris strainer captures large particles. Threaded brass neck fits 2" BSP bung opening, and funnel can be used on any steel drum or IBC.
Caution: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.
Specifications
- Ø×H:560×290 mm
- Description:Funnel, red/black
- Seq:02



