Order Entry
Export
Orders LinkContactUsLinkComponent
 

Accessories for pH electrodes

pH buffer, 4M KCl/AgCl partial gel, 150 ml
Catalog # JENW90238-60
Supplier:   Cole-Parmer
Accessories for pH electrodes
Catalog # JENW90238-60
Supplier:  Cole-Parmer
Accessories for pH electrodes
Accessories for pH electrodes

pH buffer, 4M KCl/AgCl partial gel, 150 ml

Price
84,30 €
Log in for availability
1 * 150 mL
Product Details & Documents
Specifications
  • Description:
    pH buffer, 4M KCl/AgCl partial gel, 150 ml