Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Tubes
Centrifuge Tubes
Tubes for ParasiTrap®
Tubes for ParasiTrap®
Print
Tubes for ParasiTrap®
Processing tubes II with integrated filters, Black
Catalog # 8491BLACK
Supplier:
Biosepar
Tubes for ParasiTrap®
Catalog # 8491BLACK
Supplier:
Biosepar
Expand image
10+
Return to Top
Tubes for ParasiTrap®
Processing tubes II with integrated filters, Black
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 50 Assays
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Colour:
Black
Description:
Processing tubes II with integrated filters
Seq:
01