0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Standards
Environmental Analysis Standards
ISE calibration standards
ISE calibration standards
Copper ISE standard solution, 1000 mg/l, 1000 mg/l as Cu, 500 mL
METT51344774
:
Mettler - Toledo
ISE calibration standards
METT51344774
:
Mettler - Toledo
:
ISE calibration standards
Copper ISE standard solution, 1000 mg/l, 1000 mg/l as Cu, 500 mL
:
165,00 €
1 * 500 mL
-
+
CoC_METT51344775_ISEF524D1
CoC_METT51344779_ISEN524E1
Concentration:
1000 mg/l as Cu
Pk:
500 mL
Description:
Copper ISE standard solution, 1000 mg/l