Order Entry
Export
Orders LinkContactUsLinkComponent
ISE calibration standards

ISE calibration standards

Iodide ISE standard solution, 1000 mg/l, 1000 mg/l as I, 500 mL
Catalog # METT51344776
Supplier:   Mettler - Toledo
ISE calibration standards
Catalog # METT51344776
Supplier:  Mettler - Toledo
CAS Number:  
ISE calibration standards
ISE calibration standards

Iodide ISE standard solution, 1000 mg/l, 1000 mg/l as I, 500 mL

CAS Number:  
Price
129,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/l as I
  • Pk:
    500 mL
  • Description:
    Iodide ISE standard solution, 1000 mg/l

Specifications