Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Certificates
Manufacturer Data
0
0,00 €
Order Entry
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Your Account
Export
Your Account
Orders Link
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
ISE calibration standards
ISE calibration standards
ISE calibration standards
Lead ISE standard solution, 1000 mg/l, 1000 mg/l as Pb, 500 mL
METT51344780
:
Mettler - Toledo
ISE calibration standards
METT51344780
:
Mettler - Toledo
:
Expand image
ISE calibration standards
Lead ISE standard solution, 1000 mg/l, 1000 mg/l as Pb, 500 mL
Specifications
Add to cart
Jump to:
Specifications
:
165,00 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Other Product Family Options Available
Documents
Certificates
Search by Lot Number
CoC_METT51344775_ISEF524D1
CoC_METT51344779_ISEN524E1
Specifications
Specifications
Concentration:
1000 mg/l as Pb
Pk:
500 mL
Description:
Lead ISE standard solution, 1000 mg/l
Specifications