Bottles, wide neck, PE, rectangular, with screw cap
1000 ml, 58 mm, 103×80×176 mm
Catalog # SEMA0063
Supplier: SEMADENI
Jump to:
Product Details & Documents
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
Specifications
- Capacity:1000 ml
- W×D×H:103×80×176 mm
- Neck I-Ø:58 mm



