Order Entry
Export
Orders LinkContactUsLinkComponent
Copper etchant

Copper etchant

950 mL
Catalog # 44583.WY
Corrosive
Irritant
Copper etchant
Catalog # 44583.WY
CAS Number:  
Copper etchant
Copper etchant

950 mL

CAS Number:  
Price
124,00 €
Log in for availability
1 * 950 mL
Product Details & Documents

Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

Physical Form: Liquid Density: 1.19 Appearance: Dark yellow to brown Odour: Acidic UN No.: UN3264 MDL No.: MFCD00161912

Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

Synonyms:  Copper etching solution
  • MDL Number:MFCD00161912
  • Density:1.19
  • Storage Temperature:Ambient
  • UN:3264
  • ADR:8,III
Specifications

Specifications

  • Pk:
    950 mL

Specifications