Order Entry
Export
Orders LinkContactUsLinkComponent
Fast probe master mixes

Fast probe master mixes

Fast probe master mix, 2×1 ml, 200, 2 mL
Catalog # 31005.
Supplier:   Biotium
Fast probe master mixes
Catalog # 31005.
Supplier:  Biotium
CAS Number:  
Fast probe master mixes
Fast probe master mixes

Fast probe master mix, 2×1 ml, 200, 2 mL

CAS Number:  
Price
321,00 €
Log in for availability
1 * 2 mL
Product Details & Documents

A qPCR master mix containing Cheetah™ Taq hotstart DNA polymerase, suitable for probe-based real-time PCR applications.

  • 2X probe-based qPCR master mix for fast cycling protocols
  • Suitable for all fluorescent probe types, including TaqMan®, BHQ®, and Molecular Beacon
  • Contains Cheetah™ Taq hotstart DNA polymerase

Fast Probe Master Mix is designed for fluorescent probe-based PCR applications. It is compatible with probes such as a TaqMan®, BHQ®, and Molecular Beacon. However for most PCR applications we recommend the Forget-Me-Not™ Universal Probe Master Mix, for superior activity.

Fast Probe Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.

Specifications

Specifications

  • Pk:
    2 mL
  • Description:
    Fast probe master mix, 2×1 ml
  • No. of reactions:
    200
  • Cat. No.:
    31005.
  • Application:
    qPCR probe

Specifications