Orders LinkContactUsLinkComponent
β-glucuronidase substrates
β-glucuronidase substrates
Chromogenic β-glucuronidase substrate, 1 KIT
Catalog # 10025.
Supplier:   Biotium
β-glucuronidase substrates
Catalog # 10025.
Supplier:  Biotium
CAS Number:  
β-glucuronidase substrates
β-glucuronidase substrates
Chromogenic β-glucuronidase substrate, 1 KIT
CAS Number:  
Price
371,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
β-glucuronidase substrates are easy to detect stains against the background of plant cells.
White solid soluble in water or DMSO.
Caution: Store at −20 °C and protect from light.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Chromogenic β-glucuronidase substrate
Specifications