Order Entry
Export
Orders LinkContactUsLinkComponent
Beta-galactosidase substrates

Beta-galactosidase substrates

Chromogenic β-galactosidase substrate, 1 KIT
Catalog # 10024.
Supplier:   Biotium
Beta-galactosidase substrates
Catalog # 10024.
Supplier:  Biotium
CAS Number:  
Beta-galactosidase substrates
Beta-galactosidase substrates

Chromogenic β-galactosidase substrate, 1 KIT

CAS Number:  
Price
371,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Chromogenic and fluorogenic beta-galactosidase substrates.

White solid form soluble in water and DMSO.

 : Includes 5 mg of X-gal, red-beta-D-gal, rose-beta-D-gal, purple-beta-D-gal and green-beta-D-gal.

 : Store at −20 °C and protect from light.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Chromogenic β-galactosidase substrate
  • Cat. No.:
    10024.
  • Storage conditions:
    Protect from light
  • Storage temperature:
    −20 °C

Specifications