Thioflavin T
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 2390-54-7
Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
- CAS Number:2390-54-7
- Formula:C₁₇H₁₉ClN₂S
- MDL Number:MFCD00011944
- Molecular Weight:318.86 g/mol
- Storage Temperature:Ambient
Documents
ACRO21176
SDS
Specifications
Specification Test Results
- Appearance:Yellow to golden to orange-brown Powder
- Infrared spectrum:Conforms
- Solubility:(1 mg/ml in methanol) Clear to slightly hazy yellow to yellow-green
- UV Lambda max:409 to 415 nm (10 mg/l in water)
Specifications
- Pack type:Glass bottle
- Pk:100 g



