Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, 4 position, thermostattable, for 10 mm standard cells (external water bath required)
Catalog # 634-6012
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-6012
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, 4 position, thermostattable, for 10 mm standard cells (external water bath required)
Price
515,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Cell holder, 4 position, thermostattable, for 10 mm standard cells (external water bath required)
  • Seq:
    18